2-Methoxynaphthalene ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Methyl 2-naphthyl ether

A17650.0B A17650.30
A17650.0BEA 102 EUR
A17650.0B A17650.30
2-Methoxynaphthalene ≥98%
2-Methoxynaphthalene

Purity: 98% CAS No.: 93-04-9 Molecular Formula: C11H10O Formula Weight: 158.20 Flash Point: ca 139°(282°F) Melting Point: 70-75° Boiling Point: 272-274° Beilstein Reference: 1859408 Merck Reference: 14,5997 MDL No.: MFCD00004061

Formule: C₁₁H₁₀O
Kookpunt: 272…274 °C
Smeltpunt: 70…75 °C
Dichtheid: 1,266 g/cm³ (20 °C)
Vlampunt: ∼139 °C (282 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004061
CAS nummer: 93-04-9
EINECS: 202-213-6
Merck Index: 14,5997

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR