Koper(II)oxide 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Cupric oxide, Copper oxide, Copper monoxide granulated
197440250 197445000 197440025
ACRO197440250EA 29.9 EUR
ACRO197440250 ACRO197445000 ACRO197440025
Koper(II)oxide 97%
Koper(II)oxide
Formule: CuO
MW: 79.55 g/mol
Kookpunt: 2000
Smeltpunt: 1326
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010979
CAS nummer: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 13,02674
Order Now
Specification Test Results
Appearance Black Powder
Titration Iodometric ≥96.0 %