Cadmiumchloride 99.99% (trace metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Cadmium chloride
296330050 296330250
ACRO296330050EA 50.1 EUR
ACRO296330050 ACRO296330250
Cadmiumchloride 99.99% (trace metals basis)
Cadmiumchloride
Formule: CdCl₂
MW: 183.32 g/mol
Kookpunt: 967
Smeltpunt: 568 °C
MDL Nummer: MFCD00010916
CAS nummer: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1 III
Merck Index: 12,01653
Order Now
Specification Test Results
Appearance White Powder
Total trace metal impurities ≤0.01 %