Cadmiumchloride, ultra dry ≥99.996% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Cadmium chloride
Ampuled under argon
42852.04
42852.09
42852.18
42852.30
42852.04EA
42.6
EUR
42852.04
42852.09
42852.18
42852.30
Cadmiumchloride, ultra dry ≥99.996% (metals basis)
Cadmiumchloride
Purity: 99.996%
CAS No.: 10108-64-2
Molecular Formula: CdCl2
Formula Weight: 183.32
Physical Form: Powder, Ampouled under argon
Density: 4.047
Melting Point: 568°
Boiling Point: 960°
Merck Reference: 14,1617
UN No.: 2570
MDL No.: MFCD00010916
Formule:
CdCl₂ MW: 183.32 g/mol Kookpunt: 960 °C Smeltpunt: 568 °C Dichtheid: 4.047 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010916 CAS nummer: 10108-64-2 EINECS: 233-296-7 UN: 2570 ADR: 6.1 III Merck Index: 14,1617 |