Kwikdichloride ≥99.999% (metals basis), Puratronic®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Mercuric chloride, Mercury Chloride, Mercury dichloride
10808.14
10808.22
10808.14EA
78.5
EUR
10808.14
10808.22
Kwikdichloride ≥99.999% (metals basis), Puratronic®
Kwikdichloride
Purity: 99.999%
CAS No.: 7487-94-7
Molecular Formula: Cl2Hg
Formula Weight: 271.50
Physical Form: Powder
Density: 5.4
Refractive Index: 1.8590
Appearance: White
Odour: Odourless
Melting Point: 280-282°
Boiling Point: 302°
Merck Reference: 14,5876
UN No.: 1624
MDL No.: MFCD00011041
Formule:
HgCl₂ MW: 271.5 g/mol Kookpunt: 302 °C Smeltpunt: 280…282 °C Dichtheid: 5.4 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011041 CAS nummer: 7487-94-7 EINECS: 231-299-8 UN: 1624 ADR: 6.1 II Merck Index: 14,5876 |