Zinkchloride, ultra dry ≥99.999% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
Ampuled under argon
44198.06 44198.14 44198.22
44198.06EA 89.8 EUR
44198.06 44198.14 44198.22
Zinkchloride, ultra dry ≥99.999% (metals basis)
Zinkchloride
Purity: 99.999% CAS No.: 7646-85-7 Molecular Formula: ZnCl2 Formula Weight: 136.29 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.91 Melting Point: 290° Boiling Point: 732° Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295
Formule: ZnCl₂
MW: 136.3 g/mol
Kookpunt: 732 °C
Smeltpunt: 290 °C
Dichtheid: 2.91
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011295
CAS nummer: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 14,10132