Scandium(III) chloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
18677.03
18677.06
18677.03EA
307
EUR
18677.03
18677.06
Scandium(III) chloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Scandium(III) chloride
Purity: 99.9%
CAS No.: 10361-84-9
Molecular Formula: ScCl3
Formula Weight: 151.32
Physical Form: -20 Mesh
Density: 2.39
Melting Point: 939°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,8392
MDL No.: MFCD00011221
Formule:
Cl₃Sc MW: 151.31 g/mol Smeltpunt: 939 °C Dichtheid: 2.39 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011221 CAS nummer: 10361-84-9 EINECS: 233-799-1 Merck Index: 14,8392 |