Chroom(II)chloride 99.9% (trace metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
315190050 315190010 315190250
ACRO315190050EA 200 EUR
ACRO315190050 ACRO315190010 ACRO315190250
Chroom(II)chloride 99.9% (trace metals basis)
Chroom(II)chloride
Formule: Cl₂Cr
Kookpunt: 1302 °C (1013 hPa)
Smeltpunt: 824 °C
Dichtheid: 2,87 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010947
CAS nummer: 10049-05-5
EINECS: 233-163-3
ADR: 8,III
Merck Index: 13,02264
Order Now
Specification Test Results
Appearance Off-white to grey-green Powder
Total trace metal impurities ≤0.1 %