Mangaandioxide 99% -10 mesh
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid
222581000 222585000 222580025
ACRO222581000EA 35.2 EUR
ACRO222581000 ACRO222585000 ACRO222580025
Mangaandioxide 99% -10 mesh
Mangaandioxide
Formule: MnO₂
Dichtheid: 5,03 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003463
CAS nummer: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730
Order Now
Specification Test Results
Appearance Grey to black Powder
Titration with Na2S2O3 ≥98.5 %