Palladium(II) bromide ≥99.998% (metals basis) Pd ≥ 39.5%, Premion®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Palladium dibromide, Palladium bromide
43697.03 43697.06
43697.03EA 162 EUR
43697.03 43697.06
Palladium(II) bromide ≥99.998% (metals basis) Pd ≥ 39.5%, Premion®
Palladium(II) bromide
Purity: 99.998% CAS No.: 13444-94-5 Molecular Formula: PdBr2 Formula Weight: 266.22 Physical Form: Powder Appearance: Black Melting Point: dec. MDL No.: MFCD00011170
Formule: Br₂Pd
MW: 266.23 g/mol
Smeltpunt: Dec.
Dichtheid: 5.173 g/cm³ (16 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011170
CAS nummer: 13444-94-5
EINECS: 236-588-2