Iron(II) chloride, anhydrous ≥99.5% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Iron dichloride, Ferrous chloride
31141.06
31141.18
31141.30
31141.06EA
79.1
EUR
31141.06
31141.18
31141.30
Iron(II) chloride, anhydrous ≥99.5% (metals basis)
Iron(II) chloride
Purity: 99.5%
CAS No.: 7758-94-3
Molecular Formula: Cl2Fe
Formula Weight: 126.75
Physical Form: Powder
Density: 3.160
pH: 2.5
Melting Point: 674°
Boiling Point: 1023°
Solubility: Freely soluble in water, alcohol, acetone
Merck Reference: 14,4043
UN No.: 3260
MDL No.: MFCD00011004
Formule:
Cl₂Fe MW: 126.75 g/mol Kookpunt: 1023 °C Smeltpunt: 674 Dichtheid: 3.160 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011004 CAS nummer: 7758-94-3 EINECS: 231-843-4 UN: 3260 ADR: 8 III Merck Index: 14,4043 |