Iron(III) chloride, anhydrous 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Iron trichloride, Ferric chloride
12357.09
12357.22
12357.A1
12357.A7
12357.C4
12357.09EA
48.4
EUR
12357.09
12357.22
12357.A1
12357.A7
12357.C4
Iron(III) chloride, anhydrous 98%
Iron(III) chloride
Purity: 98%
CAS No.: 7705-08-0
Molecular Formula: Cl3Fe
Formula Weight: 162.21
Physical Form: Crystalline
Density: 2.90
Appearance: Black
Odour: Odourless
pH: 1.8
Melting Point: 306°
Boiling Point: 316°
Solubility: Freely soluble in water, alcohol, ether, acetone. Slightly soluble in CS2. Insoluble in ethyl acetate
Merck Reference: 14,4019
UN No.: 1773
MDL No.: MFCD00011005
Formule:
FeCl₃ Kookpunt: 316 °C Smeltpunt: 306 °C Dichtheid: 2,90 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011005 CAS nummer: 7705-08-0 EINECS: 231-729-4 UN: 1773 ADR: 8,III Merck Index: 14,4019 |