Platinum(IV) chloride (≥57% Pt) ≥99.9% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Platinum tetrachloride
11045.03 11045.06
11045.03EA 181 EUR
11045.03 11045.06
Platinum(IV) chloride (≥57% Pt) ≥99.9% (metals basis)
Platinum(IV) chloride
Purity: 99.9% CAS No.: 13454-96-1 Molecular Formula: PtCl4 Formula Weight: 336.90 Physical Form: Crystalline Density: 4.303 Appearance: Brown Odour: Odourless Melting Point: 370° dec. UN No.: 2923 MDL No.: MFCD00011182
Formule: Cl₄Pt
MW: 336.89 g/mol
Smeltpunt: 370 °C (ontbinding)
Dichtheid: 4.303
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011182
CAS nummer: 13454-96-1
EINECS: 236-645-1
UN: 2923
ADR: 8 III