Nikkelmonoxide ≥99.998% (metals basis), Puratronic®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Nickel monoxide, Nickel oxide, Bunsenite
10819.06
10819.14
10819.22
10819.06EA
52.9
EUR
10819.06
10819.14
10819.22
Nikkelmonoxide ≥99.998% (metals basis), Puratronic®
Nikkelmonoxide
Purity: 99.998%
CAS No.: 1313-99-1
Molecular Formula: NiO
Formula Weight: 74.71
Physical Form: Powder
Density: 6.67
Appearance: Dark green
Odour: Odourless
Melting Point: 1984°
Merck Reference: 14,6512
MDL No.: MFCD00011145
Formule:
NiO MW: 74.69 g/mol Smeltpunt: 1984 °C Dichtheid: 6.67 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011145 CAS nummer: 1313-99-1 EINECS: 215-215-7 Merck Index: 14,6512 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...