Iron(II) sulphide ≥99.9% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Ferrous sulphide
14024.03 14024.09 14024.18
14024.03EA 20.6 EUR
14024.03 14024.09 14024.18
Iron(II) sulphide ≥99.9% (metals basis)
Iron(II) sulphide
Purity: 99.9% CAS No.: 1317-37-9 Molecular Formula: FeS Formula Weight: 87.91 Physical Form: -100 Mesh Powder Density: 4.74 Appearance: Brown to black Odour: Odourless Melting Point: 1195° Merck Reference: 14,4058 MDL No.: MFCD00011013
Formule: FeS
MW: 87.91 g/mol
Smeltpunt: 1195
Dichtheid: 4.74
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011013
CAS nummer: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9 III
Merck Index: 14,4058