Perylene for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.20969.0001 8.20969.0005
8.20969.0001EA 42.9 EUR
8.20969.0001 8.20969.0005
Perylene for synthesis, Sigma-Aldrich®
Perylene
Formule: C₂₀H₁₂
Kookpunt: 443 °C (1013 hPa)
Smeltpunt: 273…278 °C
Dichtheid: 1,362 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004142
CAS nummer: 198-55-0
EINECS: 205-900-9
Merck Index: 12,07325
Order Now
Specification Test Results
Solubility  - 0.0001 g/l (25 °C)
Melting Point 273 - 278 °C
Molar Mass 252.31 g/mol
Vapor Pressure  - 0.001 hPa (20 °C)