Fmoc-cys(Trt)-NovaSyn® TGA, Novabiochem®
Leverancier: Novabiochem (Part of Merck)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8.56028.0001 8.56028.0005 8.56028.0025
04-12-2656-1EA 137 EUR
04-12-2656-1 04-12-2656-5 NOVA8.56028.0025
Fmoc-cys(Trt)-NovaSyn® TGA, Novabiochem®
Sorbenten voor chromatografie Sorbenten voor SPE
The base resin is PEG-PS-copolymer (90 µm), functionalised with 4-hydroxy-methylpenoxyacetic acid.
  • Colour: White to yellow to beige
  • Form: beads
  • Store at 2 to 8 °C