VWR®, Bottle carrying systems
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
292807309 292826306 292836307 292816305 292806304
215-4172EA 123 EUR
215-4172 215-4170 215-4171 215-4169 215-4168
VWR®, Bottle carrying systems
Dragers Flessendragers
This modular bottle carrying system fits standard laboratory bottles that have a GL 45 neck finish.
  • Compatible with four bottle sizes 2, 5, 10 and 20 litres
  • All materials are fully autoclavable
  • Makes pouring liquids from the larger bottles more comfortable and safer
It is ergonomically designed and offers health and safety benefits to the user. It allows easy and safe transport of bottles around the laboratory or production facility.
Order Now
Specificaties
Environmentally Preferable