3-Isobutyl-1-methylxanthine, Millipore®
Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
410957-1 410957-250
410957-1EA 280 EUR
410957-1 410957-250
3-Isobutyl-1-methylxanthine, Millipore®
3-Isobutyl-1-methylxanthine
Formule: C₁₀H₁₄N₄O₂
Smeltpunt: 200…201 °C
MDL Nummer: MFCD00005584
CAS nummer: 28822-58-4
EINECS: 249-259-3