Theobromine

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

8963.1 8963.2
ROTH8963.1EA 0 EUR
ROTH8963.1 ROTH8963.2
Theobromine
Theobromine
Formule: C₇H₈N₄O₂
Smeltpunt: 350 °C
Dichtheid: 1,419 g/cm³ (25 °C)
MDL Nummer: MFCD00022830
CAS nummer: 83-67-0
EINECS: 201-494-2
Merck Index: 12,09418

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR