Pyridazine 98+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
131760050 131760500
ACRO131760050EA 41.6 EUR
ACRO131760050 ACRO131760500
Pyridazine 98+%
Pyridazine
Formule: C₄H₄N₂
Kookpunt: 208 °C (760 mmHg)
Smeltpunt: –8 °C
Dichtheid: 1,103 g/cm³
Vlampunt: 85 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006463
CAS nummer: 289-80-5
EINECS: 206-025-5
Merck Index: 12,08152
Order Now
Specification Test Results
Appearance Clear yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5225 to 1.5245 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.0 %