3-Amino-6-methylpyridazine
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB07039.1 BTB07039.10
MAYBBTB07039.1EA 0 EUR
MAYBBTB07039.1 MAYBBTB07039.10
3-Amino-6-methylpyridazine
3-Amino-6-methylpyridazine
Formule: C₅H₇N₃
MW: 109.13 g/mol
MDL Nummer: MFCD01569899
CAS nummer: 18591-82-7
EINECS: 242-430-3