Safety ankle boots, lace-up, Run-R 300, Heckel
Leverancier: Uvex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Run-R
UVEX6266339EA 89.7 EUR
UVEX6266339 UVEX6266338 UVEX6266340 UVEX6266342 UVEX6266341 UVEX6266344 UVEX6266343 UVEX6266346 UVEX6266345 UVEX6266337 UVEX6266348 UVEX6266347
Safety ankle boots, lace-up, Run-R 300, Heckel
Schoenen en laarzen
Metal-free, flexible and lightweight safety ankle boots made from oiled hydro nubuck leather, upper resistant to liquid penetration plus a double-desity PU2D sole, highly slip resistant and resistant to oils and hydrocarbons.
  • 3-D polyester mesh lining
  • Anatomically-shaped hygienic insole
  • Shock-absorbent heel and front
  • Metal-free protective toecap (200 J)
  • Metal-free puncture resistant midsole
Metal-free model for tradesmen, light industry, warehousing and logistics, manufacturing, dry environments, public services and humid environments.
Colour: Black/orange
Weight: 550 g
Certificaten: EN ISO 20345:2011; S3, SRC