2-Methoxy-3-nitropyridine
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SPB06058.1
SPB06058EA
SPB06058EE
MAYBSPB06058.1EA
0
EUR
MAYBSPB06058.1
MAYBSPB06058EA
MAYBSPB06058EE
2-Methoxy-3-nitropyridine
2-Methoxy-3-nitropyridine
Formule:
C₆H₆N₂O₃ Smeltpunt: 149…150 °C |
MDL Nummer:
MFCD00023459 CAS nummer: 20265-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...