2-Bromo-4-methylpyridine 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Bromo-4-picoline
350350010 350350100
ACRO350350010EA 32.8 EUR
ACRO350350010 ACRO350350100
2-Bromo-4-methylpyridine 99%
2-Bromo-4-methylpyridine
Formule: C₆H₆BrN
Kookpunt: 87 °C (10 mmHg)
Smeltpunt: 218…228 °C
Dichtheid: 1,545 g/cm³
Vlampunt: >109 °C
MDL Nummer: MFCD00082590
CAS nummer: 4926-28-7
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5600 to 1.5630 (20°C, 589 nm)
GC ≥98.5 %