2-Amino-4-methylpyridine 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Aminopicoline, 2-Amino-4-picoline, 4-Methyl-2-pyridylamine, 4-Methylpyridin-2-amine
153861000 153865000
ACRO153861000EA 39.1 EUR
ACRO153861000 ACRO153865000
2-Amino-4-methylpyridine 98%
2-Amino-4-methylpyridine
Formule: C₆H₈N₂
MW: 108.14 g/mol
Kookpunt: 230 °C (760 mmHg)
Smeltpunt: 96…101 °C
Vlampunt: 118 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006321
CAS nummer: 695-34-1
EINECS: 211-780-9
UN: 2811
ADR: 6.1 III
Merck Index: 13,00465
Order Now
Specification Test Results
Appearance Light yellow to beige to brown Flakes
Infrared spectrum Conforms
Melting point 96 to 101 °C
Water ≤0.5 %
GC ≥97.5 %