2-Benzoylpyridine 99+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
105800250 105801000
ACRO105800250EA 31.6 EUR
ACRO105800250 ACRO105801000
2-Benzoylpyridine 99+%
2-Benzoylpyridine
Formule: C₁₂H₉NO
Kookpunt: 317 °C (1013 hPa)
Smeltpunt: 42…44 °C
Dichtheid: 1,171 g/cm³ (0 °C)
Vlampunt: 150 °C
MDL Nummer: MFCD00006300
CAS nummer: 91-02-1
EINECS: 202-034-3
Order Now
Specification Test Results
Appearance Colorless to light yellow Crystalline chunks
Infrared spectrum Conforms
Melting point 42 to 44 °C
GC ≥99.0 %