Pyrrole

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

SB00554.1 SB00554.10 SB00554.50
MAYBSB00554.1EA 0 EUR
MAYBSB00554.1 MAYBSB00554.10 MAYBSB00554.50
Pyrrole
Pyrrole
Formule: C₄H₅N
Kookpunt: 130…131 °C (1013 hPa)
Smeltpunt: –24 °C
Dichtheid: 0,97 g/cm³ (20 °C)
Vlampunt: 39 °C
MDL Nummer: MFCD00005216
CAS nummer: 109-97-7
EINECS: 203-724-7
UN: 1992
ADR: 3,III
Merck Index: 12,08198

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR