2-Bromo-3-methylthiophene
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SPB08396.1
SPB08396.10
MAYBSPB08396.1EA
0
EUR
MAYBSPB08396.1
MAYBSPB08396.10
2-Bromo-3-methylthiophene
2-Bromo-3-methylthiophene
Formule:
C₅H₅BrS Kookpunt: 14…16 °C (0,3 mmHg) Dichtheid: 1,5844 g/cm³ (17.5 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00059741 CAS nummer: 14282-76-9 EINECS: 238-175-2 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...