Fluoride ISE standard solutions
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35949 29149 29153
HACH35949EA 56.5 EUR
HACH35949 HACH29149 HACH29153
Fluoride ISE standard solutions
Standaarden Anionische standaarden Fluoride standaarden
  • Traceable to NIST standard reference materials
  • Designed for use with Hach meters portfolio and other meters
  • Reliable packaging
Opgelet: 1 µg/ml = 1 mg/l