Biopsy processing/embedding cassettes, Unisette™ M406
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M406-2T M406-7T M406-2 M406-3 M406-4 M406-5 M406-6 M406-7 M406-8 M406-9 M406-10 M406-11 M406-12 M406-3T M406-4T M406-5T M406-6T M406-8T M406-9T M406-10T M406-11T M406-12T
720-2955EA 241 EUR
720-2955 SIMPM406-7T SIMPM406-2 SIMPM406-3 SIMPM406-4 SIMPM406-5 SIMPM406-6 SIMPM406-7 SIMPM406-8 SIMPM406-9 SIMPM406-10 SIMPM406-11 SIMPM406-12 720-2956 720-2957 720-2958 720-2959 SIMPM406-8T SIMPM406-9T SIMPM406-10T SIMPM406-11T SIMPM406-12T
Biopsy processing/embedding cassettes, Unisette™ M406
Histologie-cassettes Cassettes voor biopsiebehandeling
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Moulded from a special high density polymer
  • Resistant to the chemical action of most histological solvents