High performance crimping tool and jaw sets, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W225761 W225762 W225763 W225741 W225771 W225772 W225751 W225752 W225833-D W225773 W225830-C W225831-C W225832-C W225833-C W225830-D W225831-D W225832-D W225830-G W225831-G W225832-G W225833-G
WHEAW225761EA 1200 EUR
WHEAW225761 WHEAW225762 WHEAW225763 WHEAW225741 WHEAW225771 WHEAW225772 WHEAW225751 WHEAW225752 WHEAW225833-D WHEAW225773 WHEAW225830-C WHEAW225831-C WHEAW225832-C WHEAW225833-C WHEAW225830-D WHEAW225831-D WHEAW225832-D WHEAW225830-G WHEAW225831-G WHEAW225832-G WHEAW225833-G
High performance crimping tool and jaw sets, WHEATON®
Capsuleer- en decapsuleertangen
These high performance crimping tools are designed with inter-changeable jaws, so that users only need one tool, and can purchase individually sized crimpers and decappers for specific laboratory requirements.
  • Designed with external power source and cord (no battery)
  • Optional base and mounting kit
The jaw sets are available as crimper and decappers, and fit standard lab vials seals with aluminum or two-part aluminum/steel caps. The crimping tool has the ability to store upto nine programs for different caps and cap sizes. The tools are ergonomicallyMore Product Information