Electronic high power crimping tool
Leverancier: Macherey-Nagel
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MANA735700EA 1650 EUR
MANA735700
Electronic high power crimping tool
Capsuleer- en decapsuleertangen
Electronic high power crimping tool for 11 mm, 13 mm and 20 mm crimp caps.
  • Due to a more powerful motor also suitable for magnetic and bi-metal crimp caps
  • With fixed 12 V DC power supply
  • Exchangeable crimping / decapping heads
Digital LED display of crimp settings and different jaw settings can be stored in separate programs.
Certificaten: CE certificate of conformity along with one year warranty.
Bestelinformatie: Stand is available additionally.