Carbon dioxide standard solutions
Leverancier: REAGECON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SC023C SC022C SC026C SC024C
REGESC023CEA 394 EUR
REGESC023C REGESC022C REGESC026C REGESC024C
Carbon dioxide standard solutions
Standaarden Standaarden voor milieuanalyse
Standards for M965D carbon dioxide analyser, for soft drinks and brewery applications.
Order Now
Specificaties
Environmentally Preferable