Safety cans, type II AccuFlow™, galvanised steel
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7210120Z 7225130Z 7220120Z 7250130Z 7220220Z 7250230Z
119-0418EA 119 EUR
119-0418 119-0421 119-0419 119-0424 119-0420 119-0425
Safety cans, type II AccuFlow™, galvanised steel
Veiligheidscontainers
Safety cans are designed to convenient and safe storage and dispense of flammable liquids.
  • Rounded steel comfort handle protects hand injury
  • Trilingual warning label in English/Spanish/French
  • Large ID zone identifies contents to reduce misuse
Features vented AccuFlow™ manifold and Safe-Squeeze® trigger handle provides the controlled liquid flow ever and flexible metal hose of 229 mm long to reach small openings. Ergonomically designed quick-fill port with comfort lift lever makes filling easier than ever.
Fill port is equipped with a long length, long life stainless steel flame arrester which reliably dissipates heat to prevent flashback ignition. Leakproof, positive pressure relief lid automatically vents to prevent rupture or explosion and self-closes to control vapors and spills.
Certificaten: Certified by FM, UL/ULC, and TÜV and also meet OSHA and NFPA requirements, and come with a ten-year warranty.