HPLC columns, Cogent Amide™
Leverancier: MICROSOLV
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40036-05P 40036-05P-1 40036-05P-2 40036-10P 40036-10P-1 40036-10P-2 40036-15P 40036-15P-1 40036-15P-2 40036-25P 40036-P21-100 40036-P21-150 40036-P21-250 40036-P21-50 40036-P30-100 40036-P30-50 40036-SP100 40036-SP250
554-6294EA 933 EUR
554-6294 554-6295 554-6296 554-6299 554-6300 554-6301 554-6304 554-6305 554-6306 554-6309 554-6320 554-6321 554-6322 554-6323 554-6325 554-6328 554-6330 554-6331
HPLC columns, Cogent Amide™
Kolommen voor chromatografie
Cogent Amide™ has an amide functional group bonded to the silica-hydride surface direct silicon-carbon bonds. Since the ligand is attached to the surface with direct silicon carbon bonds the bonded phase does not hydrolyze in acidic conditions. Also, it does not make a strong association with acetone.
  • Excellent for sugars and other carbohydrates
This column is very stable and efficient and is recommended for reversed-phase or aqueous-normal-phase separations of biomolecules, including carbohydrates, peptides, polysaccharides or tryptic digests.
Order Now
Specificaties
Environmentally Preferable
Koolstofgehalte 2 - 3%
pH-bereik 2,5 - 7,5
Oppervlakte 390 m²/g
Temperatuurlimiet 80 °C
USP code L68