Screw caps, 9 mm, Soft-Guard™
Leverancier: MICROSOLV
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9502S-E-10C-B 9502S-E-10C-G 9502S-E-10C-K 9502S-E-10C-N 9502S-E-10C-P 9502S-E-10C-R 9502S-E-10C-Y 9502S-E-30C-B 9502S-E-30C-G 9502S-E-30C-K 9502S-E-30C-N 9502S-E-30C-P 9502S-E-30C-Y 9502S-E-34B 9502S-E-3XB 9502S-E-3XG 9502S-E-3XK 9502S-E-3XN 9502S-E-3XP 9502S-E-3XR 9502S-E-3XY
MICS9502S-E-10C-BEA 38.3 EUR
MICS9502S-E-10C-B MICS9502S-E-10C-G MICS9502S-E-10C-K MICS9502S-E-10C-N MICS9502S-E-10C-P MICS9502S-E-10C-R MICS9502S-E-10C-Y MICS9502S-E-30C-B MICS9502S-E-30C-G MICS9502S-E-30C-K MICS9502S-E-30C-N MICS9502S-E-30C-P MICS9502S-E-30C-Y MICS9502S-E-34B MICS9502S-E-3XB MICS9502S-E-3XG MICS9502S-E-3XK MICS9502S-E-3XN MICS9502S-E-3XP MICS9502S-E-3XR MICS9502S-E-3XY
Screw caps, 9 mm, Soft-Guard™
Sluitingen Sluitingen voor chromatografievials
These ribbed screw caps with septa are suitable for 12×32 mm autosampler vials.