Anti-ATRX Rabbit Polyclonal Antibody

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILP0083-C01 ILP0083-C05 ILP0083-C1
ILP0083-C01EA 319 EUR
ILP0083-C01 ILP0083-C05 VWRKILP0083-C1
Anti-ATRX Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ATRX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR