Anti-MLH1 Mouse Monoclonal Antibody [clone: ES05]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM1100-C01 ILM1100-C05 ILM1100-C1
ILM1100-C01EA 311 EUR
ILM1100-C01 ILM1100-C05 ILM1100-C1
Anti-MLH1 Mouse Monoclonal Antibody [clone: ES05]
Antilichamen
Type: Primary
Antigen: MLH1
Clonality: Monoclonal
Clone: ES05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human