Anti-ENO2 Mouse Monoclonal Antibody [clone: 500]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM0056-C01 ILM0056-C05 ILM0056-C1
ILM0056-C01EA 151 EUR
ILM0056-C01 ILM0056-C05 ILM0056-C1
Anti-ENO2 Mouse Monoclonal Antibody [clone: 500]
Antilichamen
Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: 500
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 alpha
Reactivity: Human