Anti-PAX8 Rabbit Monoclonal Antibody [clone: ZR1]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM0002-C01 ILM0002-C05 ILM0002-C1
ILM0002-C01EA 244 EUR
ILM0002-C01 ILM0002-C05 ILM0002-C1
Anti-PAX8 Rabbit Monoclonal Antibody [clone: ZR1]
Antilichamen
Type: Primary
Antigen: PAX8
Clonality: Monoclonal
Clone: ZR1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human