Anti-CR2 Mouse Monoclonal Antibody [clone: 2G9]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM8403-C01 ILM8403-C05 ILM8403-C1
ILM8403-C01EA 159 EUR
ILM8403-C01 ILM8403-C05 ILM8403-C1
Anti-CR2 Mouse Monoclonal Antibody [clone: 2G9]
Antilichamen
Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR