Anti-GATA3 Mouse Monoclonal Antibody [clone: L50-823]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM3003-C01 ILM3003-C05 ILM3003-C1
ILM3003-C01EA 173 EUR
ILM3003-C01 ILM3003-C05 ILM3003-C1
Anti-GATA3 Mouse Monoclonal Antibody [clone: L50-823]
Antilichamen
Type: Primary
Antigen: GATA3
Clonality: Monoclonal
Clone: L50-823
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR