Anti-H. pylori Rabbit Polyclonal Antibody

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILP4343-C1 ILP4343-C01 ILP4343-C05
ILP4343-C1EA 532 EUR
ILP4343-C1 ILP4343-C01 ILP4343-C05
Anti-H. pylori Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: H. pylori
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Whole Organism
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR