Anti-ACTA2 Mouse Monoclonal Antibody [clone: 1A4]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM20211-C01 ILM20211-C05 ILM20211-C1
ILM20211-C01EA 47.3 EUR
ILM20211-C01 ILM20211-C05 ILM20211-C1
Anti-ACTA2 Mouse Monoclonal Antibody [clone: 1A4]
Antilichamen
Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Baboon, Monkey, Cow, Pig, Sheep, Goat, Cat, Dog, Rabbit, Mouse, Rat, Guinea pig, Chicken