Anti-BCL2 Mouse Monoclonal Antibody [clone: 124]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM2263-C01 ILM2263-C05 ILM2263-C1
ILM2263-C01EA 63 EUR
ILM2263-C01 ILM2263-C05 ILM2263-C1
Anti-BCL2 Mouse Monoclonal Antibody [clone: 124]
Antilichamen
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR