Anti-B. burgdorferi Rabbit Polyclonal Antibody

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILP0301-C01 ILP0301-C05 ILP0301-C1
ILP0301-C01EA 101 EUR
ILP0301-C01 ILP0301-C05 ILP0301-C1
Anti-B. burgdorferi Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: B. burgodorferi
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgodorferi
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR