Anti-MME Mouse Monoclonal Antibody [clone: 56C6]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM1103-C01 ILM1103-C05 ILM1103-C1
ILM1103-C01EA 192 EUR
ILM1103-C01 ILM1103-C05 ILM1103-C1
Anti-MME Mouse Monoclonal Antibody [clone: 56C6]
Antilichamen
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: 56C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human