Anti-SDC1 Mouse Monoclonal Antibody [clone: B-A38]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM3825-C01 ILM3825-C05 ILM3825-C1
ILM3825-C01EA 81.6 EUR
ILM3825-C01 ILM3825-C05 ILM3825-C1
Anti-SDC1 Mouse Monoclonal Antibody [clone: B-A38]
Antilichamen
Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: B-A38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR