Anti-FUT4 Mouse Monoclonal Antibody [clone: MMA]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM31711-C01 ILM31711-C05 ILM31711-C1
ILM31711-C01EA 63 EUR
ILM31711-C01 ILM31711-C05 ILM31711-C1
Anti-FUT4 Mouse Monoclonal Antibody [clone: MMA]
Antilichamen
Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: MMA
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR